Learn More
Chemsplash FFP3 Fold Flat into Cup Mask with Valve
Product Code. 30058414
Change view
Click to view available options
Aerosol Type:
R - Somewhat Resistant to Oil
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Aerosol Type Unit Size
30058414 R - Somewhat Resistant to Oil Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30058414 Supplier Chemsplash Supplier No. 3528
  • ONLINE EXCLUSIVE PRICE
    329.09€
    278.42€ / Pack of 200
    Save 50.67€
    15% Off
Estimated Shipment: 07-12-2026
The FPP3 valved fold flat into cup mask has super fold flat design, 3D Structure and Large Filtration Area.
Chemsplash 3528 P3 Valved Fold Flat into Cup Mask
  • High Filtration Capacity
  • Respirator with Red Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Adjustable Nose Clip with Padded Comfort on Inside
  • Super Fold Flat Design, 3D Structure and Large Filtration Area
  • Pass the Optional Dolomite (D) Clogging Test
  • Packed Individually for Hygiene
  • Protects Against Harmful and Toxic Solid and Liquid Particles
  • No Vision Impairment and Problem-Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3528 is a premium P3 valved fold flat into cup mask. It features a super fold flat design, 3D structure, and a large filtration area. The mask is colour-coded red for ease of use. The fabric material has a high filtering efficiency and passes the optional Dolomite Clogging Test. A soft padded band rests on the nose, which is adjustable to provide an excellent fit, ensuring comfort and security. The mask protects against harmful and toxic solid and liquid particles while maintaining clear vision and speech. It is latex and graphene-free, and packed individually for hygiene.
Specifications
Aerosol Type R - Somewhat Resistant to Oil
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP3
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes