Learn More
Chemsplash FFP2 Fold Flat into Cup Mask
Description
Chemsplash 3508 FFP2 Valved Fold Flat into Cup Mask
- High Filtration Capacity
- Respirator with Yellow Color Exhalation Valve
- Exhalation Valve Reduces Hot Air Buildup
- Adjustable Nose Clip with Padded Comfort on Inside
- Woven Elasticated Adjustable Head Straps
- Super Fold Flat Design, 3D Structure and Large Filtration Area
- Pass the Optional Dolomite (D) Clogging Test
- Packed Individually for Hygiene
- Protects Against Low-Toxicity Solid and Liquid Particles
- Latex and Graphene Free
Description: The Chemsplash 3508 is a FFP2 valved fold flat into cup mask, featuring a super fold flat design, 3D structure, and a large filtration area. It is colour-coded yellow for ease of use. The mask fabric has high filtering efficiency and passes the optional Dolomite Clogging Test. An adjustable nose clip with padded comfort ensures an excellent fit, while the woven elasticated adjustable head straps offer secure and comfortable wear. This mask protects against low-toxicity solid and liquid particles and is latex and graphene-free. It is packed individually for hygiene.
Specifica
Specifica
| Aerosol Type | N - Not Resistant to Oil |
| Color | White |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP2 |
| Material | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.