missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fetuin Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| For Use With (Application) | ELISA |
| Formulation | Prepared from ammonium sulfate fractionation of bovine serum |
| Gene ID (Entrez) | 26998 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 48.4 kDa |
| Quantity | 1 g |
| Storage Requirements | Store at 4°C. After reconstitution store at -20°C. |
| Gene Alias | fetuin B, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 |
| Gene Symbol | FETUB |
| Reconstitution | Use sterile distilled water to give a solution of 1 mg/mL |
| Purity or Quality Grade | >80%, by SDS-PAGE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction