missing translation for 'onlineSavingsMsg'
Learn More

Ferrostatin 1, TRC

Product Code. 30179741
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179741 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179741 Supplier Toronto Research Chemicals Supplier No. TRCF3077005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 347174-05-4
Molecular Formula C15H22N2O2
Quantity 5 mg
Synonym 3-Amino-4-cyclohexylaminobenzoic Acid Ethyl Ester, Fer-1
InChI Formula InChI=1S/C15H22N2O2/c1-2-19-15(18)11-8-9-14(13(16)10-11)17-12-6-4-3-5-7-12/h8-10,12,17H,2-7,16H2,1H3
SMILES CCOC(=O)c1ccc(NC2CCCCC2)c(N)c1
IUPAC Name ethyl 3-amino-4-(cyclohexylamino)benzoate
Molecular Weight (g/mol) 262.35
Formula Weight 262.1681
Chemical Name or Material Ferrostatin 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.