missing translation for 'onlineSavingsMsg'
Learn More

Ferric Sulfate, MP Biomedicals

Product Code. p-3547113 Shop All MP Biomedical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 11405471

Brand: MP Biomedicals 0215804283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10028-22-5
Molecular Formula Fe2O12S3
Molecular Weight (g/mol) 399.858
InChI Key RUTXIHLAWFEWGM-UHFFFAOYSA-H
Synonym ferric sulfate, iron iii sulfate, iron persulfate, iron tersulfate, diiron tris sulphate, diiron trisulfate, ferric persulfate, ferric tersulfate, iron sesquisulfate, ferric sesquisulfate
PubChem CID 24826
ChEBI CHEBI:53438
IUPAC Name iron(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]

Specifications

Quantity 250 g
Formula Weight 399.9
Chemical Name or Material Ferric Sulfate
compliance-icons
Product Identifier
  • Ferric Sulfate Hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.