Learn More
Thermo Scientific™ X-Gluc
Product Code. 10519140 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10519140 0.5 g 500mg
10509140 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10519140 Supplier Thermo Scientific™ Supplier No. R0852
Edge
Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.
X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Recommended for:
  • Detection of GUS expression in plant cells and tissues (1)
  • Detection of infections caused by E.coli
  • Detection of bacterial contamination in food and water samples
Note:
Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.
TRUSTED_SUSTAINABILITY
Especificaciones
Detection Method Colorimetric
Product Type X-Gluc
Quantity 0.5 g
Substrate Properties Chemical Substrate
Substrate Type Beta-Glucuronidase (GUS) Substrate
For Research Use Only. Not for use in diagnostic procedures.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.