Learn More
Thermo Scientific™ MassRuler DNA Loading Dye (6X)
Product Code. 11813943 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11813943 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11813943 Supplier Thermo Scientific™ Supplier No. R0621
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X MassRuler DNA Loading Dye.
6X MassRuler DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains one dye (bromophenol blue) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solutions binds divalent metal ions and inhibits metal-dependent nucleases.
6X MassRuler DNA Loading Dye is used for conventional DNA electrophoresis (see Recommendations for use).
Highlights
  • One-color tracking of DNA migration during DNA electrophoresis 
  • No DNA masking during gel exposure to UV light
  • EDTA binds divalent metal ions and inhibits metal dependent nucleases
TRUSTED_SUSTAINABILITY
Specifications
Concentration 6X
Gel Compatibility Acrylamide Gels, Agarose Gels
Label or Dye Bromophenol Blue
Product Type DNA Loading Dye
Product Line MassRuler
Quantity 5 x 1 mL