Learn More
Thermo Scientific™ dNTP Mix (25 mM each)
Thermo Scientific™ dNTP Mix, 25mM each of dATP, dTTP, dCTP and dGTP.
Brand:  Thermo Scientific™ R1122
Product Code. 15727546
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Thermo Scientific™ dNTP Mix contains 25mM each of dATP, dTTP, dCTP and dGTP.
For direct use in PCR, reverse transcription and other applications.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
>99%
Standard PCR
Unlabeled
5 x 1 mL