Learn More
Thermo Scientific™ Biotin Chromogenic Detection Kit
Product Code. 10755651 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 Reactions
30 Reactions
Unit Size:
10 reactions
30 reactions
Product Code. 10755651
Brand: Thermo Scientific™ K0661
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific Biotin Chromogenic Detection Kit is a convenient tool for the chromogenic detection of biotinylated nucleic acid probes.
Thermo Scientific Biotin Chromogenic Detection Kit is a convenient tool for the chromogenic detection of biotinylated nucleic acid probes. The kit is optimized to reproducibly provide high sensitivity with a low background in applications, such as Southern, Northern, dot and slot blotting, and screening of viral plaques and bacterial colonies. Biotinylation of DNA/RNA probes is widely used as a safe and convenient alternative to radioactive labeling. Biotin can be incorporated into nucleic acids using various enzymatic or non-enzymatic methods, including the Biotin DecaLabel DNA Labeling Kit (Cat. No. K0651).
Features
Biotinylated probes are detected with streptavidin coupled to alkaline phosphatase (AP). Streptavidin-AP conjugates bind specifically and irreversibly to the biotin-labeled probes. The probes are then visualized using a chromogenic substrate for alkaline phosphatase BCIP/NBT, which produces a blue-purple precipitate. Visualization does not require X-ray film or other specific equipment (see Figure 1).
Highlights
High sensitivity of detection—0.03 pg of homologous DNA in dot blot hybridization with biotin-labeled probe
Low background due to optimized washing and blocking procedures
Non-radioactive detection visualization does not require X-ray film or other specific equipment (see Figure 2)
Fast and convenient—ready-to-use components
Applications
• Southern, Northern, dot/slot blots, plaque, or colony screening.
Note: This kit is optimized for hybridization procedures using nylon membranes. The detection sensitivity may vary depending on the membranes supplied by different vendors.
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Chromogenic
Format Kit
Labeling Target DNA/RNA
For Use With (Application) Southern, Northern, Dot/Slot Blots, Plaque, Colony Screening
Product Type Biotinylated Nucleic Acid Detection Kit
Quantity 10 Reactions