Learn More
Fensulfothion Oxon Sulfide, TRC
Specifications
Specifications
| Chemical Name or Material | Fensulfothion-oxon-sulfide |
| CAS | 3070-13-1 |
| Molecular Formula | C11 H17 O4 P S |
| Quantity | 250 mg |
| Synonym | Phosphoric acid, diethyl 4-(methylthio)phenyl ester, Phosphoric acid, diethyl p-(methylthio)phenyl ester (6CI,7CI,8CI), Phenol, p-(methylthio)-, diethyl phosphate (8CI), Fensulfothion oxon sulfide, NSC 132955, O,O-Diethyl O-(4-methylthiophenyl) phosphate, Fensulfothion-oxon-sulfide |
| InChI Formula | InChI=1S/C11H17O4PS/c1-4-13-16(12,14-5-2)15-10-6-8-11(17-3)9-7-10/h6-9H,4-5H2,1-3H3 |
| SMILES | CCOP(=O)(OCC)Oc1ccc(SC)cc1 |
| IUPAC Name | diethyl (4-methylsulfanylphenyl) phosphate |
| Molecular Weight (g/mol) | 276.29 |
| Formula Weight | 276.0585 |