missing translation for 'onlineSavingsMsg'
Learn More

Fenoverine, TRC

Product Code. 30399146
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399146 Supplier Toronto Research Chemicals Supplier No. TRCF248898500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenoverine
CAS 37561-27-6
Molecular Formula C26 H25 N3 O3 S
Quantity 500 mg
Synonym Fenoverine
InChI Formula InChI=1S/C26H25N3O3S/c30-26(29-20-5-1-3-7-24(20)33-25-8-4-2-6-21(25)29)17-28-13-11-27(12-14-28)16-19-9-10-22-23(15-19)32-18-31-22/h1-10,15H,11-14,16-18H2
SMILES O=C(CN1CCN(Cc2ccc3OCOc3c2)CC1)N4c5ccccc5Sc6ccccc46
IUPAC Name 2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1-phenothiazin-10-ylethanone
Molecular Weight (g/mol) 459.56
Formula Weight 459.1617
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.