missing translation for 'onlineSavingsMsg'
Learn More

Fenclorim, TRC

Product Code. 30399145
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399145 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399145 Supplier Toronto Research Chemicals Supplier No. TRCF247500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenclorim
CAS 3740-92-9
Molecular Formula C10 H6 Cl2 N2
Quantity 250 mg
Synonym 4,6-Dichloro-2-phenylpyrimidine, 2-Phenyl-4,6-dichloropyrimidine, CGA 123407, Fenclorim, NSC 27713, Pyrimidine, 4,6-dichloro-2-phenyl-
InChI Formula InChI=1S/C10H6Cl2N2/c11-8-6-9(12)14-10(13-8)7-4-2-1-3-5-7/h1-6H
SMILES Clc1cc(Cl)nc(n1)c2ccccc2
IUPAC Name 4,6-dichloro-2-phenylpyrimidine
Molecular Weight (g/mol) 225.07
Formula Weight 223.9908
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.