Learn More
Fenbuconazole D5 (phenyl D5) 100 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Fenbuconazole (phenyl D5) |
| CAS | 1398066-06-2 |
| Solvent or Matrix | Acetone |
| Molecular Formula | C19 2H5 H12 Cl N4 |
| Quantity | 1 mL |
| Synonym | Fenbuconazole (phenyl D5) |
| InChI Formula | InChI=1S/C19H17ClN4/c20-18-8-6-16(7-9-18)10-11-19(12-21,13-24-15-22-14-23-24)17-4-2-1-3-5-17/h1-9,14-15H,10-11,13H2/i1D,2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(CCc2ccc(Cl)cc2)(Cn3cncn3)C#N |
| IUPAC Name | 4-(4-chlorophenyl)-2-(2,3,4,5,6-pentadeuteriophenyl)-2-(1,2,4-triazol-1-ylmethyl)butanenitrile |
| Molecular Weight (g/mol) | 341.85 |
| Show More |