Learn More
Value Plastics Female Luer to 500 Series Barb
Product Code. 30053142 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Product Code. 30053142
Brand: Value Plastics 5012042
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/16 in.
Material Polypropylene
Packaging Quantity 25/Pk.
Inlet Female Luer x Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.