missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feed Kit B+
Click to view available options
Quantity:
2 L
Unit Size:
Each
Specifications
Specifications
| Content And Storage | • 1 x 1 L GlycanTune B+ Total Feed • 1 x 1 L EfficientFeed B+ Total Feed Store above in refrigerator (2–8°C). Protect from light and moisture. • Flash drive with manual |
| Cell Line | CHO |
| Classification | Chemically-defined |
| Form | AGT |
| Product Type | Media Supplement |
| Quantity | 2 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction