Learn More
WTW™ FDO 925 Dissolved Oxygen Sensors
Product Code. 10604635
Change view
Click to view available options
Temperature (Metric) Range:
0°C to 50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Range Unit Size
10604635 0°C to 50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10604635 Supplier WTW™ Supplier No. 201300
  • ONLINE EXCLUSIVE PRICE
    1275.00€
    1120.00€ / Pack of 1
    Save 155.00€
    12% Off
View all promotions
In Stock
Fast and flow-free, the FDO™ 925 is ideal suited for field measurement.
  • For use with field and laboratory applications with fast responding beveled membrane
  • The FDO™ 925’s small dimensions make it suitable for lab and process
  • The fast and flow-free FDO™ 925 is perfectly suited for field measurement
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±0.5% of value
Length (Metric) Cable 1.5 m
Sensor Types Dissolved Oxygen
Dissolved Oxygen (Saturation Range) 0.0 to 200%
Temperature (Metric) Range 0°C to 50°C
Includes 15m Length Cable
Diameter (Metric) Body 15.3 mm
Length (Metric) 140 mm
For Use With (Application) Field and laboratory applications with fast responding beveled membrane
Dissolved Oxygen (Concentration Range) 0.00 to 20 mg/L