missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FAU (Human) Recombinant Protein
Human FAU full-length ORF ( AAH33877, 1 a.a. - 133 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00002197-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MQLFVRAQELHTFEVTGQETVAQIKAHVASLEGIAPEDQVVLLAGAPLEDEATLGQCGVEALTTQEVAGRMLGGKVHGSLARAGKVRGQTPKVAKQEKKKKKTGRAKRRMQYNRRFVNVVPTFGKKKGPNANS
Specifications
AAH33877 | |
Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAU | |
GST |
2197 | |
Wheat germ expression system | |
10 μg | |
FAU1, FLJ22986, Fub1, Fubi, MNSFbeta, RPS30 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |