Learn More
Fasudil, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Fasudil |
| CAS | 103745-39-7 |
| Molecular Formula | C14H17N3O2S |
| Quantity | 100 mg |
| Synonym | 5-[(Hexahydro-1H-1,4-diazepin-1-yl)sulfonyl]isoquinoline, 1-(5-Isoquinolinesulfonyl)homopiperazine, AT 877, Fasudil, HA 1077, Hexahydro-1-(5-isoquinolinesulfonyl)-1H-1,4-diazepine |
| InChI Formula | InChI=1S/C14H17N3O2S/c18-20(19,17-9-2-6-15-8-10-17)14-4-1-3-12-11-16-7-5-13(12)14/h1,3-5,7,11,15H,2,6,8-10H2 |
| SMILES | O=S(C1=CC=CC2=C1C=CN=C2)(N3CCNCCC3)=O |
| IUPAC Name | 5-((1,4-diazepan-1-yl)sulfonyl)isoquinoline |
| Molecular Weight (g/mol) | 291.37 |
| Formula Weight | 291.1041 |