Learn More
Thermo Scientific™ FastRuler Middle Range DNA Ladder, Ready-to-Use, 100-5000 bp Promotion
Shop All Thermo Scientific ProductsDescription
FastRuler Middle Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
The ladder is especially useful for electrophoresis of PCR products.
Highlights
- Fast separation (8 to 14 min)
- Short separation distance (10 to 20 mm)
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use -premixed with loading dye
- Supplied with loading dye solution for sample DNA
Specifications
Specifications
| Concentration | 6X Loading Dye |
| Form | Liquid |
| Kit Contents | 6X MassRuler DNA Loading Dye, 1mL |
| Quantity | 2 x 500 μL |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.