Learn More
Epredia™ Fast Flex™ Holding Solution
Product Code. 12559946 Shop All Epredia Products
Click to view available options
Quantity:
5L
2.5 gal.
Unit Size:
5 L
Each
Product Code. 12559946
Brand: Epredia™ 9800
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2.5 gal.