Learn More
Testo™ Fast-action Surface Probe
Product Code. p-8010237
Click to view available options
Length (Metric):
80 mm
115 mm
Unit Size:
Each
Product Code. 10221013
Brand: Testo™ 0602 0393
Only null left
Add to basket
Edge
Add to basket
Specifications
Includes Fast-action Surface Probe, 5 dia. x 115mm L Probe Shaft, 12mm dia. Probe Shaft Tip, 1.2mm L Cable, –60 to 300°C Measuring Range, 3sec. Response Time, For Detection of Thermal Bridges in Walls
For Use With (Application) Detection of thermal bridges in walls
Response Time 3 sec.
Temperature Range (Metric) -60°C to +300°C
Length (Metric) 115 mm