Learn More
Testo™ Fast-action Immersion/Penetration Probe
Click to view available options
Length (Metric) Probe:
60 mm
Unit Size:
Each
Specifications
Specifications
| Includes | Immersion/Penetration Probe, 5 dia. x 60mm L Probe Shaft, 1.5 dia. x 40mm L Probe Shaft Tip, 1.2m L Cable, –60 to 800°C Measuring Range, 3 sec. Response Time, For Temperature Measurements in Pastes and Semi-solid Media, in Liquids and in the Air. |
| Certifications/Compliance | EN 60584-2 |
| For Use With (Application) | Temperature Measurements in Pastes and Semi-solid Media, in Liquids and in the Air |
| Response Time | 3 sec. |
| Temperature Range (Metric) | -60°C to +800°C |
| Length (Metric) Probe | 60 mm |