missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fas Receptor/TNFRSF6/CD95 Antibody Pair [HRP]

Product Code. 18662061 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18662061

Brand: Novus Biologicals™ NBP2795265Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Fas Receptor/TNFRSF6/CD95.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 4.7 pg/mL
Assay Sensitivity 4.69-300 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Fas Receptor/TNFRSF6/CD95.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias ALPS1A, Apo-1, APO-1 cell surface antigen, apoptosis antigen 1, apoptosis signaling receptor FAS, Apoptosis-mediating surface antigen FAS, APT1, CD95, CD95 antigen, Fas (TNF receptor superfamily, member 6), Fas AMA, FAS1, FASLG receptor, FASTM, mutant tumor necrosis receptor superfamily member 6, TNF receptor superfamily member 6, TNFRSF6, tumor necrosis factor receptor superfamily member 6, tumor necrosis factor receptor superfamily, member 6
Gene ID (Entrez) 355
Gene Symbol FAS
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 28 ng of recombinant Fas Receptor/TNFRSF6/CD95.
  • Capture Antibody: 0.3 mg/mL of mouse anti-Fas Receptor/TNFRSF6/CD95 monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-Fas Receptor/TNFRSF6/CD95 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Cancer, Cell Biology, Death Receptor Signaling Pathway, Diabetes Research, Inhibitors Activators and Regulators, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target Fas Receptor/TNFRSF6/CD95
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.3 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.