Learn More
Hellma™ Far UV Quartz Cuvette Ultra Micro Cells
Product Code. 12761695
Click to view available options
Path Length:
10 mm
Unit Size:
Each
Product Code. 12761695
Brand: Hellma™ 105203108540
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Ultra-micro Cell Cuvette
Capacity (Metric) 0.05 mL
Material Far UV Quartz
Quantity 1Each
Path Length 10 mm
Wavelength Range 200 to 2500 nm