Learn More
Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks
Product Code. p-7070791 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
1 L
2000 mL
Unit Size:
Each
Pack of 10
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 15489103
Only null left
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Constructed from borosilicate glass. Fisherbrand™ Wide Neck Conical Borosilicate Glass Erlenmeyer Flasks are graduated and available in a variety of sizes.
  • Borosilicate glass
  • Graduated
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer Neck 34 mm
Capacity (Metric) 50 mL
Material Borosilicate Glass
Height (Metric) 85 mm
Graduated Yes
Diameter (Metric) Outer 51 mm