missing translation for 'onlineSavingsMsg'
Learn More

Famciclovir, TRC

Product Code. 30396609
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396609 50 mg 50mg
30393090 100 mg -
30406637 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396609 Supplier Toronto Research Chemicals Supplier No. TRCF10112550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104227-87-4
Molecular Formula C14 H19 N5 O4
Molecular Weight (g/mol) 321.33
Synonym 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, diacetate (ester) (9CI), 9-[4-Acetoxy-3-(acetoxymethyl)but-1-yl]-2-aminopurine, 9-[4-Acetoxy-3-(acetoxymethyl)butyl]-2-aminopurine, BRL 42810, FCV, Famciclovir, Famcivir, Famcyclovir, Famtrex, Famvir, 1,3-Propanediol, 2-[2-(2-amino-9H-purin-9-yl)ethyl]-, 1,3-diacetate
IUPAC Name [2-(acetyloxymethyl)-4-(2-aminopurin-9-yl)butyl] acetate
SMILES CC(=O)OCC(CCn1cnc2cnc(N)nc12)COC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H19N5O4/c1-9(20)22-6-11(7-23-10(2)21)3-4-19-8-17-12-5-16-14(15)18-13(12)19/h5,8,11H,3-4,6-7H2,1-2H3,(H2,15,16,18)
Formula Weight 321.1437
Chemical Name or Material Famciclovir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.