Learn More
Novus Biologicals™ FAM21C 293T Cell Transient Overexpression Lysate
Product Code. 18536261 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18536261 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18536261 Supplier Novus Biologicals™ Supplier No. H00253725T01
 Restricted Item
Estimated Shipment: 17-08-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target FAM21C
Synonym Em:AC012044.3, family with sequence similarity 21, member C, KIAA0592FAM21A, VPEFVaccinia virus penetration factor, WASH complex subunit FAM21C
Gene Symbol FAM21C