Description
Additional filters are available as standard accessories. The filters, pipet holder, and nose piece are autoclavable.
Specifications
Specifications
| Material | PTFE |
| Pore Size | 0.2 μm |
| Type | Serological Pipet |
| For Use With (Equipment) | For use with pipet controller |
| Quantity | 5/Cs. |