Learn More
Tocris Bioscience™ FAI
GREENER_CHOICE
Product Code. 18724384
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724384 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18724384 Supplier Tocris Bioscience™ Supplier No. 8011/50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Reagent for RNA SHAPE-MaP experiments in live cells
FAI is a reagent for RNA SHAPE-MaP experiments in live cells. Can be used to map RNA structures in vivoby forming stable 2'-OH adducts with RNA, blocking reverse transcriptase elongation. In vitro, FAI activity can be quenched with Dithiothreitol (Cat. No. 3154) and can be used to probe the secondary structure of mouse embryonic stem cell 5S rRNA. At elevated temperatures, FAI forms more stable adducts and exhibits less hydrolysis than NAI (Cat. No 7004).
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1H-Imidazol-1-yl(2-methyl-3-furanyl)methanone
CAS 1415238-77-5
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C9H8N2O2