missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68026-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
FADS2 Monoclonal antibody specifically detects FADS2 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISASpecifications
FADS2 | |
Monoclonal | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
Mouse | |
20 μL | |
9415 | |
Store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot, ELISA | |
1G4B10 | |
Unconjugated | |
BC001449 | |
Protein A purification | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |