missing translation for 'onlineSavingsMsg'
Learn More

FADS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4B10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68026-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17711139

  • 399.00€ / 150µL
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

FADS2 Monoclonal antibody specifically detects FADS2 in Human, Mouse, Rat samples. It is validated for IHC, WB, ELISA
Specifications

Specifications

FADS2
Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Mouse
150 μL
9415
Store at -20°C
Liquid
Immunohistochemistry, Western Blot, ELISA
1G4B10
Unconjugated
BC031835
Protein A purification
Primary
Human, Mouse, Rat
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers