Learn More
Thermo Scientific™ Factor XA Protease Digestion Buffer
Product Code. 15436039
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436039 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15436039 Supplier Thermo Scientific™ Supplier No. J63878.AK
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
For His-tag protein purification from mammalian cells
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 mL
Format Liquid
Product Type Cell Lysis Reagent
For Use With (Application) For His-tag protein purification from mammalian cells
Content And Storage Ambient temperatures