Learn More
Abcam Factor Xa Activity Assay Kit (Fluorometric)
Product Code. 32113153 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 x 100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32113153 1 x 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32113153 Supplier Abcam Supplier No. ab3253581x100Tests
 Restricted Item
Estimated Shipment: 25-11-2026
Measure Factor X activity in 40m in with Factor Xa Activity Assay Kit AB325358. Sensitivity >1 ng. Use with Microplate (12 x 8 well strips).
Measure Factor X activity in 40m in with Factor Xa Activity Assay Kit AB325358. Sensitivity >1 ng. Use with Microplate (12 x 8 well strips).
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1 x 100 Tests