Learn More
uvex Faceguard for Hypervision PC/CA, Clear
Product Code. 31751178 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Material Unit Size
31751178 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31751178 Supplier uvex Supplier No. 9321318
  • ONLINE EXCLUSIVE PRICE
    18.62€
    18.18€ / Pack of 1
    Save 0.44€
    2% Off
Estimated Shipment: 24-11-2026
Faceguard model 9321318
  • Designed as a protective faceguard compatible with Hypervision PC/CA systems, this accessory ensures safety eyewear remains protected, accessible, and conveniently stored when not in use, supported by a range of practical safety accessories such as cords and cases for organized workplace use.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Autoclavable Not Autoclavable