missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Desthiobiotin, No-Weigh™ Format
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 1 mg
Unit Size:
Pack of 5
Description
- Long-chain, amine-reactive labeling reagent whose biotin-like group is elutable from streptavidin, making it ideal for purified or cell surface protein labeling and purification
- Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
- Protein labeling - tag antibodies or proteins in purified or mixed samples to enable high recovery in pull-down assays with streptavidin beads
- Cell surface labeling - modifies only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
Specifications
Specifications
| Product Type | Sulfo-NHS-LC-Desthiobiotin |
| Quantity | 5 x 1 mg |
| Content And Storage | Store desiccated at -20°C. |
| Labeling Method | Chemical Labeling |
| Shipping Condition | Wet Ice |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction