Learn More
Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent the loss of reagent reactivity and contamination over time by minimizing the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
- Protein labeling: Biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
- Amine-reactive: Reacts with primary amines (-NH2), such as side-chain of lysines (K) or amino-termini of polypeptides
- PEGylated: Spacer arm contains hydrophilic, 12-unit, polyethylene glycol (PEG) group
- Enhances solubility: PEGylation imparts water solubility to biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
- Irreversible: Forms permanent amide bonds; spacer arm cannot be cleaved
- Very long reach: Spacer arm (total length added to target) is 56A0; this reduces steric hindrance when binding to avidin molecules
Specifications
Specifications
| Quantity | 10 x 1 mg |
| Content And Storage | Store desiccated at -5 to -30°C. |
| Shipping Condition | Room Temperature |
| Product Line | EZ-Link, No-Weigh |
| Solubility | Water |
| Format | Solid |
| Label Type | Biotin & Analogs |