Learn More
behr Labor-Technik™ behrotest™extractor 100 ml with stopcock / EZ100H
Product Code. 10288030
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10288030 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10288030 Supplier behr Labor-Technik™ Supplier No. B00217970
Extract a desired compound with limited solubility in a solvent. behr Labor-Technik™ EZ 100/HAHN Soxhlet Extractor recycles a small amount of solvent to dissolve a larger amount of material.
  • For 100mL extractions
  • Includes spigot
Specifications
Product Type Soxhlet Extractor
Capacity (Metric) 100 mL
Includes Spigot