Learn More
Motic™ Eyepiece Adapter, Moticam
Product Code. 15405222
Click to view available options
Diameter (Metric):
OD: 30mm
OD: 38mm
Unit Size:
Each
Product Code. 15405222
Brand: Motic™ 1101002301181
Only null left
Add to basket
Add to basket
Select the most appropriate adapter to connect your Moticam to a microscope eyepiece. Moticam™ Eyepiece Adapter, Moticam facilitates digital documentation of your findings.
  • For use with Moticam camera.
  • Fits directly onto eyepiece.
Specifications
Type Eyepiece Adapter
Diameter (Metric) OD: 38mm
For Use With (Equipment) Moticam