Learn More
Karl Hecht Extraction attachments 30ml
Brand:  Karl Hecht 41155006
Extraction attachments 30ml
Product Code. 10023761
  • ONLINE EXCLUSIVE PRICE
    92.40€
    63.21€ / Each
    Save 29.19€
    31% Off
Only null left
Estimated Shipment: 19-02-2026