Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Allihn Condenser
Product Code. p-7222478
Click to view available options
Capacity (Metric):
30 mL
70 mL
150 mL
200 mL
Unit Size:
Each
Product Code. 10204631
Brand: Lenz Laborglasintrumente 05365046
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor in compact and robust design
  • With flask connection
  • Made of DURAN™ tubing
  • Allihn condenser with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 200 mL
Material Duran Glass
Standard Taper Joint NS 45/40