Learn More
Lenz Laborglasintrumente Extraction Thimbles Porosity 0
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05405243
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10050241
  • 55.07€ / Each
Estimated Shipment: 06-09-2024
Description
  • Extraction thimbles
  • Glass insert with bores for solids
  • Made of DURAN™ tubing
Specifications
Specifications
150 mL
Borosilicate Glass (3.3)
0