Learn More
Gestigkeit Harry Series heating bath, 2 units, 1L, 1000W, 230V, relay output
Brand:  Gestigkeit Harry SMR2
Series heating bath, 2 units, 1L, 1000W, 230V, relay output
Product Code. 10009100
  • ONLINE EXCLUSIVE PRICE
    1307.25€
    934.58€ / Pack of 1
    Save 372.67€
    28% Off
Estimated Shipment: 06-11-2026