Learn More
Miele Professional™ Pump ADP 86 with External Booster Pump
Product Code. 30533231 Shop All Miele Products
Change view
Click to view available options
Product Type:
Booster Pump
Unit Size:
1 sheet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
30533231 Booster Pump 1 sheet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30533231 Supplier Miele Professional™ Supplier No. 12711800
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
ADP 86 for unpressurised demineralised water
  • To compensate for low DI water pressure on site
  • ADP 86 with external booster pump for unpressurised demineralised water
TRUSTED_SUSTAINABILITY
Specifications
Product Type Booster Pump
For Use With (Equipment) Washer-Disinfectors
Depth (English) 11 13/16 in.
Depth (Metric) 30.00 cm
Dimensions (L x W x H) 30.00 x 18.00 x 23.50 cm
Height (English) 9 5/16 in.
Height (Metric) 23.50 cm
Length (English) 11 13/16 in.
Length (Metric) 30.00 cm
Width (English) 7 1/8 in.
Width (Metric) 18.00 cm
Electrical Requirements 200/230 VAC, 50 Hz
Frequency 50 Hz
Quantity 1 Ea.