Learn More
Memmert™ Extension of DAkkS Factory Calibration for SN450plus
Shop All Memmert Products
Click to view available options
Unit Size:
Each
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
Specifications
| For Use With (Equipment) | SN450plus Sterilizer |
| Product Type | Factory Calibration Extension |