Learn More
Memmert™ Extension of DAkkS Factory Calibration for SF450plus
Product Code. 17528295 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF450plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17528295 SF450plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17528295 Supplier Memmert™ Supplier No. SF450plusE39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) SF450plus Sterilizer
Product Type Factory Calibration Extension