Learn More
Memmert™ Extension of DAkkS Factory Calibration for SF110plus
Product Code. 17528115 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SF110plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17528115 SF110plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17528115 Supplier Memmert™ Supplier No. SF110plusE39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Sterilizers.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) SF110plus Sterilizer
Product Type Factory Calibration Extension