Learn More
Memmert™ Extension of DAkkS Factory Calibration for IN110plus
Product Code. 17577405 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17577405 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17577405 Supplier Memmert™ Supplier No. IN110plusE39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Incubators.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IN110plus Incubator
Product Type Factory Calibration Extension