Learn More
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar
Product Code. 11788509
Change view
Click to view available options
Capacity (Metric):
8 L
10 L
20 L
34 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11788509 34 L Each
11768509 10 L Each
11778509 20 L Each
11758509 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11788509 Supplier Taylor Wharton™ Supplier No. 34XTB11
  • ONLINE EXCLUSIVE PRICE
    1780.00€
    1660.00€ / Pack of 1
    Save 120.00€
    6% Off
 Restricted Item
Estimated Shipment: 02-02-2027
The durable ribbed high-strength aluminium body, and superior vacuum performance and insulation provide versatile extended-time storage.
  • Provides extended-time storage
  • High-strength aluminium body
  • Provides maximum holding times with superior vacuum performnce and insulation
Specifications
Description Long-term Cyrogenice Storage Dewar
Model 34XTB11
Capacity (Metric) 34 L
Material Aluminum
For Use With (Application) Cryogenic storage