Learn More
Taylor Wharton™ XT Series Long-Term Cryogenic Storage Dewar
Product Code. 11758509
Click to view available options
Capacity (Metric):
8 L
10 L
20 L
34 L
Unit Size:
Each
Product Code. 11758509
Brand: Taylor Wharton™ XTL8
Only null left
Add to basket
Add to basket
The durable ribbed high-strength aluminium body, and superior vacuum performance and insulation provide versatile extended-time storage.
  • Provides extended-time storage
  • High-strength aluminium body
  • Provides maximum holding times with superior vacuum performnce and insulation
Specifications
Description Long-term Cyrogenice Storage Dewar
Model XTL8
Capacity (Metric) 8 L
Material Aluminum
For Use With (Application) Cryogenic storage