Learn More
Wheaton™ Extended Controlled Dropper Tip
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric):
13 mm (Tip)
20 mm (Tip)
8 mm (Tip)
Color:
Natural
Purple
White
Yellow
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Yellow |
| Material | LDPE |
| Drop Size | 41 μL |
| Product Type | Extended Controlled Dropper Tip |
| Diameter (Metric) | 8 mm (Tip) |